missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Testosterone |
| CAS | 58-22-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H28 O2 |
| Quantity | 25 g |
| Synonym | 17beta-Hydroxyandrost-4-en-3-one, Testosterone, Testosterone Enantate Imp. D (EP), Testosterone Propionate Imp. C (EP), Testosterone Isocaproate Imp. A (EP), Androst-4-en-3-one','17-hydroxy-','(17β)-, Testosterone (7CI,8CI), (17β)-17-Hydroxyandrost-4-en-3-one, (+)-Testosterone, 17β-Hydroxy-Δ4-androsten-3-one, 17β-Hydroxyandrost-4-en-3-one, 17β-Hydroxyandrost-4-ene-3-one, 17β-Testosterone, 4-Androsten-3-one-17β-ol, AA 2500, Andro 100, AndroForte 5, AndroGel, Androderm, Androlin, Andronaq, Andropatch, Androst-4-en-17β-ol-3-one, Androst-4-ene-17β-ol-3-one, Andrusol, Axiron, CDB 111C, COL 1621, CP 601B, Compleo, Cristerona T, Geno-cristaux Gremy, Glowmin, Homosteron, Homosterone, Intrinsa, Mertestate, NSC 9700, Nasobol, Neotestis, Noseafix, Opterone, Oreton, Orquisteron, Perandren, Percutacrine androgenique, Primotest, Primoteston, Relibra, Striant, Synandrol F, Teslen, Testandrone, Testiculosterone, Testim, Testobase, Testoderm, Testogel, Testolent, Testolin, Testopatch, Testopel, Testopropon, Testosteroid, Testosteron, Testoviron Schering, Testoviron T, Testro AQ, Testrone, Testryl, Tostrelle, Tostrex, Viatrel, Virormone, Virosterone, Δ4-Androsten-17β-ol-3-one |
| InChI Formula | InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC Name | (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 288.42 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu