Learn More
Bachem 25GR Fmoc-Gly-Wang resin (200-400 mesh, 0.5-0.8 mmol/g)
Brand:  Bachem 4003223.0025
Encompass_Preferred
25GR Fmoc-Gly-Wang resin (200-400 mesh, 0.5-0.8 mmol/g)
Product Code. 16612566
  • £617.01 / 25g