missing translation for 'onlineSavingsMsg'
Learn More

Polysulfide Oil Blank in Mineral Oil, VHG™

Product Code. 17903166
Click to view available options
Quantity:
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
This item is not returnable. View return policy

Product Code. 17903166

Brand: VHG Labs VHGPS13MBLK250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Polysulfide Oil Blank in Mineral Oil
TRUSTED_SUSTAINABILITY

Specifications

Methods XRF
Shipping Condition Room Temperature
Solvent or Matrix 13 cSt Mineral Oil
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.