Learn More
Melford X-GlcA Sodium Salt
Product Code. 30018099
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
Product Code. 30018099
Brand: Melford B721000.25
Histochemical substrate for β-D-Glucuronidase.
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative histochemical marker of specific GUS expression in cells and tissue. X-GlcA is cleaved by GUS at the B1 glucuronic bond between glucuronic acid and the 5-Bromo-4-chloro-3-indolyl part of X-GlcA via hydrolysis. The cleavage of X-GlcA results in the precipitation of water insoluble blue dichloro-dibromo-indigo precipitate at the site of enzymatic cleavage. Allow to warm to room temperature before using.
Specifications
Quantity 250 mg