missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verbascoside, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Verbascoside |
| CAS | 61276-17-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C29 H36 O15 |
| Quantity | 250 mg |
| Synonym | β-D-Glucopyranoside','2-(3,4-dihydroxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-','4-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoate], β-D-Glucopyranoside','2-(3,4-dihydroxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-','4-[3-(3,4-dihydroxyphenyl)-2-propenoate]','(E)-, (3,4-Dihydroxyphenyl)ethyl O-α-rhamnopyranosyl(1→3)-4-O-caffeoyl-β-D glucopyranoside, Acteoside, Distinctive Active Powder SL Special, Distinctive Phytostem Lilac, Kusaginin, NSC 603831, O'''-Desarabinosyllavandulifolioside A, Russetinol, Stereospermin, TJC 160, Verbascoside, trans-Acteoside, trans-Verbascoside |
| InChI Formula | InChI=1S/C29H36O15/c1-13-22(36)23(37)24(38)29(41-13)44-27-25(39)28(40-9-8-15-3-6-17(32)19(34)11-15)42-20(12-30)26(27)43-21(35)7-4-14-2-5-16(31)18(33)10-14/h2-7,10-11,13,20,22-34,36-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27+,28+,29-/m0/s1 |
| SMILES | C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO)[C@H]2OC(=O)\C=C\c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | [(2R,3R,4R,5R,6R)-6-[2-(3,4-dihydroxyphenyl)ethoxy]-5-hydroxy-2-(hydroxymethyl)-4-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl] (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 624.59 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction