missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valsartan N2-Glucuronide, TRC
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
Specifications
Specifications
| Chemical Name or Material | Valsartan N2-Glucuronide |
| Analyte or Component Names | Valsartan N2-Glucuronide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H37 N5 O9 |
| InChI Formula | InChI=1S/C30H37N5O9/c1-4-5-10-21(36)34(22(16(2)3)29(40)41)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)27-31-33-35(32-27)28-25(39)23(37)24(38)26(44-28)30(42)43/h6-9,11-14,16,22-26,28,37-39H,4-5,10,15H2,1-3H3,(H,40,41)(H,42,43)/t22-,23-,24-,25+,26-,28+/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn(n3)[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)[C@@H](C(C)C)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[5-[2-[4-[[[(1S)-1-carboxy-2-methylpropyl]-pentanoylamino]methyl]phenyl]phenyl]tetrazol-2-yl]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 611.643 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction