missing translation for 'onlineSavingsMsg'
Learn More

Tiotropium Bromide Monohydrate, Mikromol™

Product Code. 17960516
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17960516

Brand: Mikromol MM1293.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tiotropium Bromide Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiotropium bromide hydrate
CAS 139404-48-1
Analyte or Component Names Tiotropium bromide hydrate
Shipping Condition Room Temperature
Molecular Formula C19 H22 N O4 S2 . Br . H2 O
Certifications/Compliance ISO 17034
Synonym Tiotropium Bromide Monohydrate, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., Tiotropium bromide hydrate, (1alpha,2beta,4beta,5alpha,7beta)-7-[(Hydroxydi-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide hydrate
InChI Formula InChI=1S/C19H22NO4S2.BrH.H2O/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H;1H2/q+1;;/p-1/t11-,12-,13+,16-,17+;;
SMILES O.[Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5
IUPAC Name (1R,2R,4S,5S,7s)-7-(2-hydroxy-2,2-di(thiophen-2-yl)acetoxy)-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-9-ium bromide hydrate
Molecular Weight (g/mol) 490.43
Quantity 250 mg
Formula Weight 489.0279 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.