missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Benzoate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Testosterone Benzoate
Specifications
Specifications
| Chemical Name or Material | Testosterone Benzoate |
| CAS | 2088-71-3 |
| Analyte or Component Names | Testosterone Benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H32 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Testosterone benzoate, Testosterone, benzoate (7CI,8CI), NSC 524278, Testosterone 17-benzoate, Androst-4-en-3-one, 17-(benzoyloxy)-, (17beta)-, (17beta)-17-(Benzoyloxy)androst-4-en-3-one, 17beta-Benzoyloxyandrost-4-en-3-one, 17beta-Hydroxyandrost-4-en-3-one benzoate, Androst-4-en-17beta-ol-3-one benzoate |
| InChI Formula | InChI=1S/C26H32O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h3-7,16,20-23H,8-15H2,1-2H3/t20-,21-,22-,23-,25-,26-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)c5ccccc5 |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] benzoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction