missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synephrine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Synephrine
Specifications
Specifications
| Chemical Name or Material | Synephrine |
| CAS | 94-07-5 |
| Analyte or Component Names | Synephrine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, Benzyl alcohol, p-hydroxy-α-[(methylamino)methyl]- (8CI), (±)-N-Methyloctopamine, (±)-S38537-9, (±)-Synephrine, 1-(4-Hydroxyphenyl)-2-(methylamino)ethanol, 1-(4-Hydroxyphenyl)-N-methylethanolamine, 1-Hydroxy-4-[(1-hydroxy-2-methylamino)ethyl]benzene, Advantra Z, Analeptin, DL-Synephrine, Ethaphene, N-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]-N-methylamine, NSC 166285, NSC 170956, Oxedrine, Parasympatol, S 38537-9, Simpalon, Simpatol, Sympaethamin, Sympaethamine, Sympathol, Synephrin, Synephrine, Synthenate, dl-1-(4-Hydroxyphenyl)-2-(methylamino)ethanol, p-Hydroxy-α-[(methylamino)methyl]benzyl alcohol, β-Methylamino-α-(4-hydroxyphenyl)ethyl alcohol |
| InChI Formula | InChI=1S/C9H13NO2/c1-10-6-9(12)7-2-4-8(11)5-3-7/h2-5,9-12H,6H2,1H3 |
| SMILES | CNCC(O)c1ccc(O)cc1 |
| IUPAC Name | 4-[1-hydroxy-2-(methylamino)ethyl]phenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction