missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NTERA-2 Whole Cell Lysate
Shop All Bio Techne Products
Click to view available options
:
0.25 mg
Specifications
Specifications
| Preparation Method | This lysate is prepared as a positive control for separation by SDS-PAGE and subsequent Western blot analysis. Lysates are prepared in denaturing buffer WITHOUT reducing agents (i.e. 2-mercaptoethanol (BME) or dithiothreitol (DTT)). If reducing conditions are desired, add a reducing agent prior to heating. Heat lysate to 95 degrees C for 5 minutes and rapidly cool. The recommended loading amount is 0.01-0.03 mg per lane. |
| Quantity | 0.25 mg |
| Content And Storage | Store at -80C. Avoid freeze-thaw cycles. |
| Target | NTERA-2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction