missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyoscine Butylbromide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Hyoscine Butylbromide
Specifications
Specifications
| Chemical Name or Material | Hyoscine Butylbromide |
| CAS | 149-64-4 |
| Analyte or Component Names | Hyoscine Butylbromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H30 N O4 . Br |
| Certifications/Compliance | ISO 17034 |
| Synonym | Hyoscine Butylbromide, N-Butylscopolammonium bromide, (-)-Scopolamine butylbromide, Amisepan, Buscapine, Buscol, Buscolamin, Buscolysin, Buscolysine, Buscopan, Butylmin, Butylscopolamine bromide, Butylscopolammonium bromide, Donopon, Hyoscine butobromide, Hyoscine butyl bromide, Joscine, Monospan, N-Butylhyoscine bromide, N-Butylhyoscinium bromide, N-Butylscopolamine bromide, N-Butylscopolaminium bromide, Scobro, Scobron, Scobutil, Scobutyl, Scopolamine bromobutylate, Scopolamine butobromide, Scopolamine butyl bromide, Scopolamine-N-butyl bromide, Scopolan, Sparicon, Sporamin, Sporamin (pharmaceutical), Stibron, Tirantil, (1alpha,2beta,4beta,5alpha,7beta)-9-Butyl-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide (1:1), 8-Butyl-6beta,7beta-epoxy-3alpha-hydroxy-1alphaH,5alphaH-tropanium bromide (-)-tropate, [7(S)-(1alpha,2beta,4beta,5alpha,7beta)]-9-Butyl-7-(3-hydroxy-1-oxo-2-phenylpropoxy)-9-methyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide, (-)-Tropic acid ester with 8-butyl-6beta,7beta-epoxy-3alpha-hydroxy-1alphaH,5alphaH-tropanium bromide |
| InChI Formula | InChI=1S/C21H30NO4.BrH/c1-3-4-10-22(2)17-11-15(12-18(22)20-19(17)26-20)25-21(24)16(13-23)14-8-6-5-7-9-14;/h5-9,15-20,23H,3-4,10-13H2,1-2H3;1H/q+1;/p-1/t15-,16-,17-,18+,19-,20+,22+;/m1./s1 |
| SMILES | [Br-].CCCC[N@@+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4 |
| IUPAC Name | (1α,2β,4β,5α,7β)-9-Butyl-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction