missing translation for 'onlineSavingsMsg'
Learn More

Drostanolone Propionate, Mikromol™

Product Code. 17924276
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17924276 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17924276 Supplier Mikromol Supplier No. MM3574.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Drostanolone Propionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Drostanolone Propionate
CAS 521-12-0
Analyte or Component Names Drostanolone Propionate
Shipping Condition Room Temperature
Molecular Formula C23 H36 O3
Certifications/Compliance ISO 17025
Synonym 2MDTP, 32379, Drolban, Dromostanolone propionate, Drostanolone propionate, Emdisterone, MDHT, Masterid, Masteril, Masteron, Masterone, Medrotestron propionate, Medrotestrone propanoate, Medrotestrone propionate, Permastril, Prometholone, (2alpha,5alpha,17beta)-2-Methyl-17-(1-oxopropoxy)-androstan-3-one, 5alpha-Androstan-3-one, 17beta-hydroxy-2alpha-methyl-, propionate (6CI,7CI,8CI), 17beta-Hydroxy-2alpha-methyl-5alpha-androstan-3-one propionate, 17beta-Hydroxy-2alpha-methylandrostan-3-one propionate, 2alpha-Methyl-17beta-hydroxy-5alpha-androstan-3-one 17-propionate, 2alpha-Methyl-17beta-propionoxy-5alpha-androstan-3-one, 2alpha-Methyl-4,5-dihydrotestosterone propionate, 2alpha-Methylandrostan-17beta-ol-3-one propionate, 2alpha-Methyldihydrotestosterone propionate
InChI Formula InChI=1S/C23H36O3/c1-5-21(25)26-20-9-8-17-16-7-6-15-12-19(24)14(2)13-23(15,4)18(16)10-11-22(17,20)3/h14-18,20H,5-13H2,1-4H3/t14-,15+,16+,17+,18+,20+,22+,23+/m1/s1
SMILES CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)[C@H](C)C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(2R,5S,8R,9S,10S,13S,14S,17S)-2,10,13-trimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 360.53
Quantity 250 mg
Formula Weight 360.2664 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.