missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-D-alanine Methyl Ester Hydrochloride, TRC

Product Code. 30132947
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132947 250 mg 250mg
30134306 2.5 g 2.5g
30007055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132947 Supplier Toronto Research Chemicals Supplier No. TRCC363525250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112346-82-4
Formula Weight 173.001
InChI Formula InChI=1S/C4H8ClNO2.ClH/c1-8-4(7)3(6)2-5;/h3H,2,6H2,1H3;1H/t3-;/m1./s1
IUPAC Name methyl (2S)-2-amino-3-chloropropanoate;hydrochloride
Molecular Formula C4H8ClNO2.HCl
Molecular Weight (g/mol) 174.02
SMILES Cl.COC(=O)[C@H](N)CCl
Synonym (S)-Methyl 2-Amino-3-chloropropanoate Hydrochloride, Cycloserine Impurity 3
Chemical Name or Material 3-Chloro-D-alanine Methyl Ester Hydrochloride

Specifications

Quantity 250 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.