missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylthioxanthone (2-ITX), Mikromol™

Product Code. 30049585
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30049585

Brand: Mikromol MM2585.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Isopropylthioxanthone (2-ITX)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Isopropylthioxantone ITX
CAS 5495-84-1
Analyte or Component Names 2-Isopropylthioxantone ITX
Shipping Condition Room Temperature
Molecular Formula C16 H14 O S
Certifications/Compliance ISO 17034
Synonym 2-(1-Methylethyl)-9H-thioxanthen-9-one, Thioxanthen-9-one, 2-isopropyl- (7CI,8CI), 2-(1-Methylethyl)-9H-thioxanthen-9-one, 2-ITX, 2-Isopropyl-9H-thioxanthen-9-one, 2-Isopropylthioxanthen-9-one, 2-Isopropylthioxanthenone, 2-Isopropylthioxanthone, ITX, JRCure ITX, Omnirad ITX, Photocure ITX-P, Quantacure 597, Quantacure 659, Quantacure ITX, Speedcure ITX
InChI Formula InChI=1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3
SMILES CC(C)c1ccc2Sc3ccccc3C(=O)c2c1
IUPAC Name 2-propan-2-ylthioxanthen-9-one
Molecular Weight (g/mol) 254.35
Quantity 250 mg
Formula Weight 254.0765 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.