missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1’-Thiobismethane-d3 (Dimethyl Sulfide-d6), TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Dimethyl-d6 sulfide |
| CAS | 926-09-0 |
| Molecular Formula | C2 2H6 S |
| Quantity | 250 mg |
| Synonym | 1,1'-Thiobismethane-d3 |
| InChI Formula | InChI=1S/C2H6S/c1-3-2/h1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])SC([2H])([2H])[2H] |
| IUPAC Name | trideuterio(trideuteriomethylsulfanyl)methane |
| Molecular Weight (g/mol) | 68.17 |
| Formula Weight | 68.0567 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction