Learn More
BTX™ 25-Well Plate Seal
Description
BTX Multi-well Electroporation plate sealing films are used to:
- Prevent well cross-contamination during electroporation
- Mark off wells that have been used
- Maintain sterility in unused wells
Specifications
Specifications
| Color | Clear |
| Autoclavable | Not Autoclavable |
| No. of Wells | 24 |
| Adhesive | Yes |
| Pierceable | Yes |
| Plate Compatibility | BTX 25-well Electroporation Plates |
| Fit for Purpose | Sterile |