missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TNF-alpha Hyperactive Mutant Protein

Product Code. 18757576
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18757576

Brand: R&D Systems™ 8599TA025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mutant form with approximately 5-fold greater activity

  • Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D.
  • ED50 for this effect is 20 to 100 pg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01375
For Use With (Application) Bioactivity
Gene ID (Entrez) 7124
Molecular Weight (g/mol) 17 kDa
Name APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Quantity 25 μg
Source E. coli-derived human TNF-alpha protein Lys87-Leu233 (Leu233Phe), with an N-terminal Met-Arg-Lys-Arg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol TNF
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.