missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-37/IL-1F7 Protein

Product Code. 18746269
Click to view available options
:
Recombinant Human IL-37/IL-1F7 Protein
This item is not returnable. View return policy

Product Code. 18746269

Brand: R&D Systems™ 9225IL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-37/IL-1F7 Protein is derived from E. coli. The Recombinant Human IL-37/IL-1F7 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZH6-2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein.
Gene ID (Entrez) 27178
Molecular Weight (g/mol) 21 kDa
Name FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (z
Quantity 25μg
Source E. coli-derived human IL-37/IL-1F7 protein Ser2-Asp192
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity Measured by its ability to inhibit LPS-stimulated IL-8 secretion by HK-2 human kidney epithelial cells. The ED50 for this effect is 0.25-1.25 μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.