missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CUL5/NEDD8/RBX2 Complex Protein, CF
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Specifications
Specifications
| Accession Number | Q93034, Q9UBF6, Q15843 |
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT |
| Molecular Weight (g/mol) | Theroretical: 103 kDa (neddylated CUL5), 13 kDa (RB x 2) |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL5/NEDD8/RBX2 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie Blue stain. |
| Protein | CUL5/NEDD8/RBX2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction