missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prostaglandin E Synthase 2/PTGES2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7627-SP
This item is not returnable.
View return policy
Description
Prostaglandin E Synthase 2/PTGES2 Monoclonal specifically detects Prostaglandin E Synthase 2/PTGES2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Prostaglandin E Synthase 2/PTGES2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C9orf15, chromosome 9 open reading frame 15, EC 5.3.99.3, FLJ14038, gamma-interferon-activated transcriptional element-binding factor 1, GATE-binding factor 1, GBF1, membrane-associated prostaglandin E synthase 2, Microsomal prostaglandin E synthase 2, microsomal prostaglandin E synthase-2, mPGES2, mPGES-2, PGES2, PGES2MGC11289, prostaglandin E synthase 2 | |
| E. coli-derived recombinant Human Prostaglandin E Synthase 2/PTGES2, Glu88-His377, Accession No. Q9H7Z7 | |
| 25 μg | |
| 80142.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 998012 | |
| Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL | |
| Q9H7Z7 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects Human Prostaglandin E Synthase 2/PTGES2 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction