missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB48941-SP
This item is not returnable.
View return policy
Description
EMR2 Monoclonal antibody specifically detects EMR2 in Human samples. It is validated for Flow Cytometry
Specifications
| EMR2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2 | |
| NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry | |
| 494014 | |
| Flow Cytometry 0.25 ug/106 cells | |
| AAI27006 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 30817 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction