missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 6/ANGPTL6 Antibody, R&D Systems™

Description
Angiopoietin-like Protein 6/ANGPTL6 Monoclonal antibody specifically detects Angiopoietin-like Protein 6/ANGPTL6 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Angiopoietin-like Protein 6/ANGPTL6 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 856606 |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q8ROZ6 |
| Gene Alias | AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?