missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK-7 Antibody, R&D Systems™

Sheep Antibody
Brand: R&D Systems AF7749-SP
This item is not returnable.
View return policy
Description
ALK-7/Activin Receptor Type 1C specifically detects ALK-7/Activin Receptor Type 1C in Human samples. It is validated for Immunohistochemistry.
Specifications
| ALK-7/Activin Receptor Type 1C | |
| Polyclonal | |
| Immunohistochemistry 10-25 μg/mL | |
| Q8NER5 | |
| Sheep | |
| Antigen Affinity-purified | |
| Primary | |
| Detects human ALK-7 in direct ELISAs. | |
| Human | |
| Lyophilized |
| Immunohistochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ALK-7, Met1-Glu113, Accession No. Q8NER5 | |
| 25 μg | |
| 130399.0 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction