Learn More
Hanna Instruments™ 25 Tests Reagent Set For Ammonia in Fresh Water
Click to view available options
Range:
0.0 to 2.5mg/L
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | For Ammonia in Fresh Water |
| Includes | With Protective Cap, Batteries and Instructions |
| No. of Tests | 25 |
| Quantity | 1 Set |
| Range | 0.0 to 2.5mg/L |
| Test Method | Nessler |
| Type | Reagent Set |