Learn More
Invitrogen™ 25-OH Vitamin D3 Recombinant Rabbit Monoclonal Antibody (RM3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA527880
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
25-OH Vitamin D3 (calcifediol) is a prohormone produced by the liver by hydroxylation of vitamin D3 and is used to measure vitamin D status and detect itamin D deficiency.
Specifications
| 25-OH Vitamin D3 | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| RM3 | |
| Unconjugated | |
| 25-hydroxy vitamin D; 25-hydroxyvitamin D; 25-OH Vitamin D | |
| 25-OH Vitamin D3 BSA conjugate. | |
| 100 μL | |
| Primary | |
| -20°C | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.