missing translation for 'onlineSavingsMsg'
Learn More
Learn More
25-OH Vitamin D3 Rabbit, Clone:RM3, Abnova™
Rabbit monoclonal antibody raised against 25-OH Vitamin D3.
Brand: Abnova MAB16909.100uL
This item is not returnable.
View return policy
Specifications
| 25-OH Vitamin D3 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| 25-OH Vitamin D3 BSA conjugate. | |
| This antibody reacts to 25-OH Vitamin D3. It does not cross-react with Vitamin D3, Vitamin D2, and 25-OH Vitamin D2. | |
| Antibody | |
| IgG |
| ELISA | |
| RM3 | |
| Competitive ELISA (0.5ng/mL-1.5ng/mL) The optimal working dilution should be determined by the end user. |
|
| Rabbit | |
| 100 μL | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction