missing translation for 'onlineSavingsMsg'
Learn More
Learn More
25-OH Vitamin D3 Antibody-antigen Complex Antibody (RM428), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26039B
This item is not returnable.
View return policy
Description
25-OH Vitamin D3 Antibody-antigen Complex Monoclonal antibody specifically detects 25-OH Vitamin D3 Antibody-antigen Complex in Nonspecies specific samples. It is validated for ELISA
Specifications
| 25-OH Vitamin D3 Antibody-antigen Complex | |
| Monoclonal | |
| Biotin | |
| PBS | |
| 25-OH Vitamin D3 Antibody-antigen Complex | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| RM428 | |
| ELISA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction