missing translation for 'onlineSavingsMsg'
Learn More

25-O-Deacetyl Rifabutin, TRC

Product Code. 30354166
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354166 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354166 Supplier Toronto Research Chemicals Supplier No. TRCD19898025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 25-O-Deacetyl Rifabutin
CAS 100324-63-8
Purity Grade Notes HPLC
Molecular Formula C44 H60 N4 O10
Quantity 25 mg
Synonym 16-Deacetylrifabutin, Spiro[9,4-(epoxypentadeca[1,11,13]trienimino)-2H-furo[2',3':7,8]naphth[1,2-d]imidazole-2,4'-piperidine]-5,10,26(3H,9H)-trione, 6,16,18,20-tetrahydroxy-14-methoxy-7,9,15,17,19,21,25-heptamethyl-1'-(2-methylpropyl)-, (9S,12E,14S,15S,16S,17R,18R,19R,20S,21S,22E,24Z)-, Rifamycin XIV, 25-O-deacetyl-1',4-didehydro-1-deoxy-1,4-dihydro-5'-(2-methylpropyl)-1-oxo-, (9S,12E,14S,15S,16S,17R,18R,19R,20S,21S,22E,24Z)-6,16,18,20-Tetrahydroxy-14-methoxy-7,9,15,17,19,21,25-heptamethyl-1'-(2-methylpropyl)spiro[9,4-(epoxypentadeca[1,11,13]trienimino)-2H-furo[2',3':7,8]naphth[1,2-d]imidazole-2,4'-piperidine]-5,10,26(3H,9H)-trione, 16-Deacetyl rifabutin, 25-Deacetyl rifabutin, 25-Hydroxy-rifabutin, 25-O-Deacetylrifabutin, 25-O-Desacetylrifabutin, Antibiotic LM 565, LM 565
InChI Formula InChI=1S/C44H60N4O10/c1-21(2)20-48-17-15-44(16-18-48)46-32-29-30-38(52)27(8)40-31(29)41(54)43(9,58-40)57-19-14-28(56-10)24(5)36(50)26(7)37(51)25(6)35(49)22(3)12-11-13-23(4)42(55)45-34(39(30)53)33(32)47-44/h11-14,19,21-22,24-26,28,35-37,47,49-52H,15-18,20H2,1-10H3,(H,45,55)/b12-11+,19-14+,23-13-/t22-,24+,25+,26-,28-,35-,36+,37+,43?/m0/s1
SMILES CO[C@H]1\C=C\OC2(C)Oc3c(C)c(O)c4C(=O)C(=C5NC6(CCN(CC(C)C)CC6)N=C5c4c3C2=O)NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@@H]1C)\C
Molecular Weight (g/mol) 804.97
Formula Weight 804.4309
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.