missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ OUL 35
GREENER_CHOICE

Product Code. 18779926
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18779926

Brand: Tocris Bioscience™ 6344/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective PARP-10 inhibitor

OUL 35 is a selective PARP-10 (also known as ADP-ribosyltransferase Diptheria Toxin-like 10, ARTD10) inhibitor (IC50 = 0.329 μM). Displays selectivity for PARP-10 over a panel of PARP family members. Rescues HeLa cells from PARP-10-induced cell death. Sensitizes cancer cells to hydroxyurea-induced genotoxic stress.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6336-34-1
Synonym 4,4′-Oxybis[benzamide]
Quantity 25 mg
Molecular Formula C14H12N2O3
Formula Weight 256.26
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.