Learn More
25 LT DIMETHYLFORMAMIDE FOR PEPTIDE SYNTHESIS,plastic drum 25L
Brand: Fisher Chemical D/3848/25
1344.25 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
25 LT DIMETHYLFORMAMIDE FOR PEPTIDE SYNTHESIS,plastic drum 25L
Code : 22
Additional Details : CAS Number : 68-12-2 Weight : 23.60000kg Transport : UN number : 2265 Chem class : 3 Pack group : III