missing translation for 'onlineSavingsMsg'
Learn More

25-Hydroxyvitamin D3 3-Hemisuccinate, TRC

Product Code. 30398496
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398496 5 mg -
30399457 1 mg -
30394683 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398496 Supplier Toronto Research Chemicals Supplier No. TRCH9958555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69511-19-9
Molecular Formula C31 H48 O5
Molecular Weight (g/mol) 500.71
Synonym (3β,5Z,7E)- 3-(Hydrogen Butanedioate)-9,10-Secocholesta-5,7,10(19)-triene-3,25-diol, 25-Hydroxycholecalciferol-3-hemisuccinate, Calcidiol 3-Hemisuccinate
IUPAC Name 4-[(1S,3Z)-3-[(2E)-2-[(7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexyl]oxy-4-oxobutanoic acid
SMILES C[C@H](CCCC(C)(C)O)C1CCC2\C(=C\C=C/3\C[C@H](CCC3=C)OC(=O)CCC(=O)O)\CCC[C@]12C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C31H48O5/c1-21-10-13-25(36-29(34)17-16-28(32)33)20-24(21)12-11-23-9-7-19-31(5)26(14-15-27(23)31)22(2)8-6-18-30(3,4)35/h11-12,22,25-27,35H,1,6-10,13-20H2,2-5H3,(H,32,33)/b23-11+,24-12-/t22-,25+,26?,27?,31-/m1/s1
Formula Weight 500.3502
Chemical Name or Material 25-Hydroxyvitamin D3 3-Hemisuccinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.