missing translation for 'onlineSavingsMsg'
Learn More
Learn More
25-Hydroxyvitamin D3 3-Glucuronide (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 25-Hydroxyvitamin D3 3-Glucuronide |
| CAS | 170512-14-8 |
| Molecular Formula | C33 H52 O8 |
| Quantity | 1 mg |
| Synonym | (3β,5Z,7E)-25-Hydroxy-9,10-secocholesta-5,7,10(19)-trien-3-yl β-D-Glucopyranosiduronic Acid |
| InChI Formula | InChI=1S/C33H52O8/c1-19-10-13-23(40-31-28(36)26(34)27(35)29(41-31)30(37)38)18-22(19)12-11-21-9-7-17-33(5)24(14-15-25(21)33)20(2)8-6-16-32(3,4)39/h11-12,20,23-29,31,34-36,39H,1,6-10,13-18H2,2-5H3,(H,37,38)/b21-11+,22-12-/t20-,23+,24-,25+,26+,27+,28-,29+,31-,33-/m1/s1 |
| SMILES | C=C1CC[C@H](O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)C/C1=C/C=C3[C@@](CC[C@]4([H])[C@H](C)CCCC(O)(C)C)([H])[C@]4(C)CCC/3 |
| IUPAC Name | (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(((S,Z)-3-(2-((1R,3aS,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexyl)oxy)tetrahydro-2H-pyran-2-carboxylic acid |
| Molecular Weight (g/mol) | 576.76 |
| Formula Weight | 576.37 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?