missing translation for 'onlineSavingsMsg'
Learn More

25-Desacetyl Rifapentin, TRC

Product Code. 30357617
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357617 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357617 Supplier Toronto Research Chemicals Supplier No. TRCD28872025MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 25-Desacetyl Rifapentin
CAS 79039-56-8
Molecular Formula C45 H62 N4 O11
Quantity 25 mg
Synonym 25-Desacetylrifapentine, Rifamycin, 3-[[(4-cyclopentyl-1-piperazinyl)imino]methyl]-25-O-deacetyl-, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., 3-[[(4-Cyclopentyl-1-piperazinyl)imino]methyl]-25-O-deacetylrifamycin, 25-Deacetylrifapentine, 25-Desacetyl rifapentin, L 14583
InChI Formula InChI=1S/C45H62N4O11/c1-23-12-11-13-24(2)44(57)47-35-30(22-46-49-19-17-48(18-20-49)29-14-9-10-15-29)40(54)32-33(41(35)55)39(53)28(6)42-34(32)43(56)45(7,60-42)59-21-16-31(58-8)25(3)37(51)27(5)38(52)26(4)36(23)50/h11-13,16,21-23,25-27,29,31,36-38,50-55H,9-10,14-15,17-20H2,1-8H3,(H,47,57)/b12-11+,21-16+,24-13-,46-22+/t23-,25+,26+,27-,31-,36-,37+,38+,45?/m0/s1
SMILES CO[C@H]1\C=C\OC2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@@H]1C)\C)c(\C=N\N5CCN(CC5)C6CCCC6)c(O)c4c3C2=O
Molecular Weight (g/mol) 834.99
Formula Weight 834.4415
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.