missing translation for 'onlineSavingsMsg'
Learn More
Learn More
24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone, TRC
Specifications
Specifications
| Chemical Name or Material | 24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone |
| Analyte or Component Names | 24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C31 H48 O7 |
| Synonym | (24S)-24,25-Dihydro-24,25-dihydroxyfusidic Acid 21,24-Lactone, Sodium Fusidate Impurity D |
| InChI Formula | InChI=1S/C31H48O7/c1-16-19-10-13-30(6)26(29(19,5)12-11-21(16)33)22(34)14-20-25(23(37-17(2)32)15-31(20,30)7)18-8-9-24(28(3,4)36)38-27(18)35/h16,19-24,26,33-34,36H,8-15H2,1-7H3/b25-18-/t16-,19?,20+,21+,22-,23+,24+,26?,29+,30+,31+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@H]1[C@@H](O)CC[C@@]2(C)C1CC[C@@]3(C)C2[C@H](O)C[C@H]4\C(=C\5/CC[C@H](OC5=O)C(C)(C)O)\[C@H](C[C@]34C)OC(=O)C |
| IUPAC Name | [(3S,4R,8S,10S,11R,13R,14S,16S,17Z)-3,11-dihydroxy-17-[(6S)-6-(1-hydroxy-1-methyl-ethyl)-2-oxo-tetrahydropyran-3-ylidene]-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-16-yl] acetate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?