missing translation for 'onlineSavingsMsg'
Learn More

24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone, TRC

Product Code. 30372560
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372560 1 mg 1mg
30386230 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372560 Supplier Toronto Research Chemicals Supplier No. TRCD4535951MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone
Analyte or Component Names 24S,25-Dihydroxy Fusidic Acid Delta-Valerolactone
Shipping Condition Room Temperature
Molecular Formula C31 H48 O7
Synonym (24S)-24,25-Dihydro-24,25-dihydroxyfusidic Acid 21,24-Lactone, Sodium Fusidate Impurity D
InChI Formula InChI=1S/C31H48O7/c1-16-19-10-13-30(6)26(29(19,5)12-11-21(16)33)22(34)14-20-25(23(37-17(2)32)15-31(20,30)7)18-8-9-24(28(3,4)36)38-27(18)35/h16,19-24,26,33-34,36H,8-15H2,1-7H3/b25-18-/t16-,19?,20+,21+,22-,23+,24+,26?,29+,30+,31+/m1/s1
Solution Type Neat
SMILES C[C@H]1[C@@H](O)CC[C@@]2(C)C1CC[C@@]3(C)C2[C@H](O)C[C@H]4\C(=C\5/CC[C@H](OC5=O)C(C)(C)O)\[C@H](C[C@]34C)OC(=O)C
IUPAC Name [(3S,4R,8S,10S,11R,13R,14S,16S,17Z)-3,11-dihydroxy-17-[(6S)-6-(1-hydroxy-1-methyl-ethyl)-2-oxo-tetrahydropyran-3-ylidene]-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-16-yl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 532.71
Quantity 1 mg
Formula Weight 532.34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.