missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(24S)-24,25-Dihydroxyvitamin D3, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
(24S)-24,25-Dihydroxyvitamin D3 ((24S)-24,25-Dihydroxycholecalciferol) is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs.
Specifications
Specifications
| Chemical Name or Material | (24S)-24,25-Dihydroxyvitamin D3 |
| CAS | 55700-58-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H44O3 |
| Quantity | 5 mg |
| Synonym | (24S)-24,25-Dihydroxycholecalciferol |
| SMILES | C=C1/C(C[C@@H](O)CC1)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CC[C@H](O)C(C)(C)O)CC3 |
| Molecular Weight (g/mol) | 416.64 |
| Formula Weight | 416.64 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction