missing translation for 'onlineSavingsMsg'
Learn More

(24S)-24,25-Dihydroxyvitamin D3, MedChemExpress

Product Code. 16471519
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16471519

Brand: Medchem Express HY15439

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (24S)-24,25-Dihydroxyvitamin D3 ((24S)-24,25-Dihydroxycholecalciferol) is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs.

(24S)-24,25-Dihydroxyvitamin D3 ((24S)-24,25-Dihydroxycholecalciferol) is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs.

Specifications

Chemical Name or Material (24S)-24,25-Dihydroxyvitamin D3
CAS 55700-58-8
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 1 mg
Synonym (24S)-24,25-Dihydroxycholecalciferol
SMILES C=C1/C(C[C@@H](O)CC1)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CC[C@H](O)C(C)(C)O)CC3
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 98.99%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.