missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 245 mm x 245 mm Square Treated Tissue Culture Dishes

Product Code. p-7217810
Encompass_Preferred
Click to view available options
Quantity:
16 Dishes/Cs.
4 Dishes/Pk.
Unit Size:
Pack of 16
Pack of 4
This item is not returnable. View return policy

Product Code. 17148061

Brand: STEMCELL Technologies 27140

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intended for culturing anchorage-dependent cells

  • Tissue culture dishes, 245 mm x 245 mm, intended for culturing anchorage-dependent cells

Specifications

For Use With (Application) Cell Culture
Quantity 4 Dishes/Pk.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.