missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 245 mm x 245 mm Square Treated Tissue Culture Dishes
Description
- Tissue culture dishes, 245 mm x 245 mm, intended for culturing anchorage-dependent cells
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 4 Dishes/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?