missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 24-well Insert 2in1, PC-Membrane
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
£238.85 - £370.00
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
16164947
|
BRAND™
782700 |
0.4 μm | 48 x 1 |
£333.00
Pack of 48 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
16174947
|
BRAND™
782701 |
0.4 μm | 4 x 12 |
£301.00
Pack of 48 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
16184947
|
BRAND™
782706 |
8 μm | 48 x 1 |
£370.00
Pack of 48 |
In Stock
Log in to see stock. |
||||
16194947
|
BRAND™
782707 |
8 μm | 4 x 12 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Use Inserts in a hanging or standing position
- For 24-well plates or culture dishes
- Achieve uniform and controlled cell growth due to surface treatment, without coatings
Specifications
Polycarbonate | |
Sterile | |
24 |