Learn More
Hamilton™ 24 gauge, Kel-F Hub Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Click to view available options
Needle Gauge:
24 G
Unit Size:
Pack of 6
Description
Specification Sheet
- Gauge – 24 gauge
- Point Style – 2, 3, or 4
- Needle Length – Custom Needle Length
- Autoclavable – Yes
- Gas Sterilizable – Yes
- Sales Quantity – 6 needles per pack
- Hub – Kel-F
Specifications
Specifications
| Needle Gauge | 24 G |
| Product Type | Kel-F Hub Needles |