missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 36 cm

Product Code. 16365322 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 Array
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16365322

Brand: Applied Biosystems™ 4404687

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use for fragment analysis and rapid sequencing applications.

The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Capillary Length 36 cm
Content And Storage Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
Description 24-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, Human Identification Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
For Use With (Equipment) 3500xL Genetic Analyzer
For Use With (Application) Sequencing, Human Identification
Product Type 3500xL Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.