missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 36 cm

Product Code. 15859836 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15859836 1 Array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15859836 Supplier Applied Biosystems™ Supplier No. 4404687
Only null left
Add to basket
Add to basket

Use for fragment analysis and rapid sequencing applications.

The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Capillary Length 36 cm
Content And Storage Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
Description 24-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, Human Identification Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
For Use With (Equipment) 3500xL Genetic Analyzer
For Use With (Application) Sequencing, Human Identification
Product Type 3500xL Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.