missing translation for 'onlineSavingsMsg'
Learn More

δ23-FK-506(>90%), TRC

Product Code. 30136447
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136447 25 mg 25mg
30006827 5 mg 5mg
30007776 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136447 Supplier Toronto Research Chemicals Supplier No. TRCF37001525MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104987-16-8
Molecular Formula C44H67NO11
Molecular Weight (g/mol) 786
Synonym 15,19-Epoxy(5E,9E)-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone, 8,11,12,13,14,15,16,17,18,19,24,25,26,26a-tetradecahydro-19-hydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propen-1-yl)-, (3S,4R,8R,12S,14S,15R,16S,18R,19R,26aS)-, 15,19-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone, 8,11,12,13,14,15,16,17,18,19,24,25,26,26a-tetradecahydro-19-hydroxy-3-[2-(4-hydroxy-3-methoxycyclohexyl)-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propenyl)-, [3S-[3R*[E(1S*,3S*,4S*)],4S*,5E,8S*,9E,12R*,14R*,15S*,16R*,18S*,19S*,26aR*]]- (9CI), (3S,4R,8R,12S,14S,15R,16S,18R,19R,26aS)-8,11,12,13,14,15,16,17,18,19,24,25,26,26a-Tetradecahydro-19-hydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propen-1-yl)-15,19-epoxy(5E,9E)-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone, δ23-FK 506, delta23-FK-506
SMILES CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)\[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(=C\[C@@H](CC=C)C(=O)\C=C\[C@H]2C)\C)OC)[C@H](C[C@H]4C)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C44H67NO11/c1-10-13-32-21-26(2)20-27(3)22-37(53-8)40-38(54-9)24-30(6)44(51,56-40)41(48)42(49)45-19-12-11-14-33(45)43(50)55-39(28(4)15-17-34(32)46)29(5)23-31-16-18-35(47)36(25-31)52-7/h10,15,17,21,23,27-28,30-33,35-40,47,51H,1,11-14,16,18-20,22,24-25H2,2-9H3/b17-15+,26-21+,29-23+/t27-,28+,30+,31-,32+,33-,35+,36+,37-,38-,39-,40+,44+/m0/s1
Formula Weight 785.4714
Chemical Name or Material 23,24-Anhydrotacrolimus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.