missing translation for 'onlineSavingsMsg'
Learn More

22-Keto-21-thia-3,6,9-trioxatricosan-1-ol, TRC

Product Code. 30394602
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394602 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394602 Supplier Toronto Research Chemicals Supplier No. TRCK31500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 22-Keto-21-thia-3,6,9-trioxatricosan-1-ol
CAS 130727-50-3
Molecular Formula C26 H52 O5 S
Quantity 25 mg
InChI Formula InChI=1S/C26H52O5S/c1-2-3-4-5-10-13-16-23-32-24-17-14-11-8-6-7-9-12-15-18-29-19-20-30-21-22-31-26(28)25-27/h27H,2-25H2,1H3
SMILES CCCCCCCCCSCCCCCCCCCCCOCCOCCOC(=O)CO
IUPAC Name 2-[2-(11-nonylsulfanylundecoxy)ethoxy]ethyl 2-hydroxyacetate
Molecular Weight (g/mol) 476.75
Formula Weight 476.3535
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.