missing translation for 'onlineSavingsMsg'
Learn More

22-Hydroxy Mifepristone, TRC

Product Code. 30138772
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138772 10 mg 10mg
30007140 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138772 Supplier Toronto Research Chemicals Supplier No. TRCH94844510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105012-15-5
Molecular Formula C29 H35 N O3
Molecular Weight (g/mol) 445.59
Synonym Estra-4,9-dien-3-one, 11-[4-(dimethylamino)phenyl]-17-hydroxy-17-(3-hydroxy-1-propyn-1-yl)-, (11β,17β)-, Estra-4,9-dien-3-one, 11-[4-(dimethylamino)phenyl]-17-hydroxy-17-(3-hydroxy-1-propynyl)-, (11β,17β)- (9CI), (11β,17β)-11-[4-(Dimethylamino)phenyl]-17-hydroxy-17-(3-hydroxy-1-propyn-1-yl)estra-4,9-dien-3-one, RU 42698, 22-Hydroxy Mifepristone, 22-Hydroxymifepristone, Hydroxymifepristone
IUPAC Name (8S,11R,13S,14S,17S)-11-[4-(dimethylamino)phenyl]-17-hydroxy-17-(3-hydroxyprop-1-ynyl)-13-methyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CN(C)c1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@@]3(O)C#CCO)[C@@H]4CCC5=CC(=O)CCC5=C24

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H35NO3/c1-28-18-25(19-5-8-21(9-6-19)30(2)3)27-23-12-10-22(32)17-20(23)7-11-24(27)26(28)13-15-29(28,33)14-4-16-31/h5-6,8-9,17,24-26,31,33H,7,10-13,15-16,18H2,1-3H3/t24-,25+,26-,28-,29-/m0/s1
Formula Weight 445.2617
Chemical Name or Material 22-Hydroxy Mifepristone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.