missing translation for 'onlineSavingsMsg'
Learn More

22-Dehydro Cholesterol-d7 3-Acetate, TRC

Product Code. 30357798
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357798 10 mg 10mg
30358554 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30357798 Supplier Toronto Research Chemicals Supplier No. TRCD22947710MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 22-Dehydro Cholesterol-d7 3-Acetate
Molecular Formula C29 D7 H39 O2
Quantity 10 mg
Synonym 5,22E-Cholestadien-3beta-ol-d7 Acetate, 5,22E-Cholestadien-3beta-yl-d7 Acetate, (3beta,22E)-Cholesta-5,22-dien-3-ol-d7 3-Acetate
InChI Formula InChI=1S/C29H46O2/c1-19(2)8-7-9-20(3)25-12-13-26-24-11-10-22-18-23(31-21(4)30)14-16-28(22,5)27(24)15-17-29(25,26)6/h7,9-10,19-20,23-27H,8,11-18H2,1-6H3/b9-7+/t20-,23+,24+,25?,26+,27+,28+,29-/m1/s1/i1D3,2D3,19D
SMILES [2H]C([2H])([2H])C([2H])(C\C=C\[C@@H](C)C1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)C)C([2H])([2H])[2H]
IUPAC Name [(3S,8S,9S,10R,13R,14S)-10,13-dimethyl-17-[(E,2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)hept-3-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molecular Weight (g/mol) 433.717
Formula Weight 433.394
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.