missing translation for 'onlineSavingsMsg'
Learn More
Learn More
22,23-Dihydro-25-cyclohexylavermectin B1, TRC
Specifications
Specifications
| Chemical Name or Material | 22,23-Dihydro-25-cyclohexylavermectin B1 |
| CAS | 142680-85-1 |
| Molecular Formula | C50H76O14 |
| Quantity | 50 mg |
| Synonym | 25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)-22,23-dihydroavermectin A1a, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran] Avermectin A1a Derivative, 22,23-Dihydro Doramectin |
| InChI Formula | InChI=1S/C50H76O14/c1-27-13-12-16-34-26-57-47-42(51)30(4)21-37(50(34,47)54)48(53)60-36-22-35(63-49(25-36)20-19-29(3)45(64-49)33-14-10-9-11-15-33)18-17-28(2)44(27)61-41-24-39(56-8)46(32(6)59-41)62-40-23-38(55-7)43(52)31(5)58-40/h12-13,16-17,21,27,29,31-33,35-47,51-52,54H,9-11,14-15,18-20,22-26H2,1-8H3/b13-12+,28-17+,34-16+/t27-,29-,31-,32-,35+,36-,37-,38-,39-,40-,41-,42+,43-,44-,45-,46-,47+,49+,50+/m0/s1 |
| SMILES | CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]3[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O[C@H]6C[C@@H](C\C=C\3/C)O[C@@]7(CC[C@H](C)[C@H](O7)C8CCCCC8)C6)[C@]45O)C)O[C@@H](C)[C@@H]1O |
| Molecular Weight (g/mol) | 901.13 |
| Formula Weight | 900.5235 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?