missing translation for 'onlineSavingsMsg'
Learn More
Learn More
21-O-Acetyl Dexamethasone, TRC
Specifications
Specifications
| Chemical Name or Material | Dexamethasone Acetate |
| CAS | 1177-87-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H31 F O6 |
| Quantity | 100 mg |
| Synonym | Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-acetate (6CI,8CI), (11β,16α)-21-(Acetyloxy)-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, 16α-Methyl-9α-fluoroprednisolone 21-acetate, Dalalone DP, Dalalone LA, Decadron LA, Decadronal, Dectancyl, Dex-Cortidelt acetate, Dexa-Cortisyl, Dexamethasone 21-acetate, Dexamethasone acetate, Fortecortin, Fortecortin (crystal suspension), NSC 39471 |
| InChI Formula | InChI=1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Molecular Weight (g/mol) | 434.5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?