missing translation for 'onlineSavingsMsg'
Learn More

21-Hydroxypregnenolone, TRC

Product Code. 30400770
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400770 250 mg -
30395608 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400770 Supplier Toronto Research Chemicals Supplier No. TRCH952470250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1164-98-3
Molecular Formula C21H32O3
Molecular Weight (g/mol) 332.48
Synonym 3beta,21-Dihydroxy-5-pregnen-20-one, 5-Pregnen-3beta,21-diol-20-one, NSC 60793, Pregn-5-ene-3beta,21-diol-20-one
IUPAC Name 2-hydroxy-1-[(3S,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMILES C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H32O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h3,14-18,22-23H,4-12H2,1-2H3/t14-,15-,16-,17-,18+,20-,21-/m0/s1
Formula Weight 332.2351
Chemical Name or Material 21-Hydroxypregnenolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.